3-Methyl-2-furoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methylfuran-2-carboxylic acid

A204364-1G A204364-5G A204364-25G A204364-250MG
103817-548EA 19 USD
103817-548 103848-208 77086-374 77086-376 77302-908 77685-519
3-Methyl-2-furoic acid 98%
3-Methyl-2-furoic acid

Precaución: for Research Use Only

Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 236.4 °C (760 mm Hg)
Melting Pt: 133…137 °C
Flash Pt: 96.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014111
CAS Number: 4412-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR