3-Methylenecyclobutanecarbonitrile 97%

Supplier: Ambeed
Danger

A178616-10G A178616-1G A178616-25G A178616-5G
103807-360EA 10 USD
103807-360 103845-698 77029-340 77083-134 77083-136 77083-138
3-Methylenecyclobutanecarbonitrile 97%
3-Methylenecyclobutanecarbonitrile

Precaución: for Research Use Only

Formula: C₆H₇N
MW: 93.13 g/mol
Boiling Pt: 171.7 °C (760 mm Hg)
Flash Pt: 58.3 °C
Storage Temperature: Freezer
MDL Number: MFCD01085401
CAS Number: 15760-35-7
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR