ɑ-Methyl-γ-butyrolactone 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Methyldihydrofuran-2(3H)-one, Dihydro-3-methyl-2(3H)-furanone, α-Methyl-γ-butyrolactone
A196779-100G
A196779-25G
A196779-5G
A196779-1G
103816-864EA
10
USD
103816-864
103847-478
76691-076
77085-450
77085-452
77302-526
ɑ-Methyl-γ-butyrolactone 95%
alpha-Methyl-gamma-butyrolactone
Precaución: for Research Use Only
Formula:
C₅H₈O₂ Boiling Pt: 197 °C (760 mm Hg) Flash Pt: 75 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005396 CAS Number: 1679-47-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...