3-Methylcrotonic acid 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
3,3-Dimethylacrylic acid, 3-Methylbut-2-enoic acid
A100226-100G
A100226-10G
A100226-500G
A100226-1000G
A100226-25G
103807-754EA
126
USD
103807-754
76970-114
76970-116
76970-118
77296-598
77548-862
3-Methylcrotonic acid 97%
3-Methylcrotonic acid
Precaución: for Research Use Only
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 194.5 °C (760 mm Hg) Melting Pt: 65…70 °C Flash Pt: 93.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004366 CAS Number: 541-47-9 UN: 3261 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...