3-Methylbenzofuran 98%

Supplier: Ambeed
Danger

A632741-1G A632741-25G A632741-10G A632741-250MG A632741-5G
103840-288EA 109 USD
103840-288 76844-466 76844-468 77280-418 77280-420 77308-016
3-Methylbenzofuran 98%
3-Methylbenzofuran

Precaución: for Research Use Only

Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 194.8 °C (760 mm Hg)
Melting Pt: 19 °C (760 mm Hg)
Flash Pt: 63.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00205641
CAS Number: 21535-97-7
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR