3-Methylpyrocatechol 95%

Supplier: Ambeed
Warning

Synonyms: 3-Methylcatechol, 3-Methylbenzene-1,2-diol

A106400-100G A106400-10G A106400-25G A106400-1G A106400-5G
103797-032EA 10 USD
103797-032 103822-786 77270-558 77270-560 77270-562 77283-476 77309-642
3-Methylpyrocatechol 95%
3-Methylpyrocatechol

Precaución: for Research Use Only

Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 240.9 °C (760 mm Hg)
Melting Pt: 62…68 °C
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00016435
CAS Number: 488-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR