2,3-Diaminotoluene 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methylbenzene-1,2-diamine, Toluene-2,3-diamine, 1,2-Diamino-3-methylbenzene

A358547-25G A358547-100G A358547-10G A358547-500G A358547-1G A358547-5G
103826-212EA 5 USD
103826-212 103837-218 76840-380 77106-882 77106-884 77106-886 77306-692 77306-694
2,3-Diaminotoluene 97%
2,3-Diaminotoluene

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 255 °C (760 mm Hg)
Melting Pt: 59…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00011589
CAS Number: 2687-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR