3-Methylazetidine hydrochloride 95%

Supplier: Ambeed
Warning

A172992-10G A172992-250MG A172992-1G
103806-776EA 43 USD
103806-776 103845-068 77029-158 77029-160 77082-294
3-Methylazetidine hydrochloride 95%
3-Methylazetidine hydrochloride

Precaución: for Research Use Only

Formula: C4H10ClN
Storage Temperature: Refrigerator
MDL Number: MFCD09743448
CAS Number: 935669-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR