3-Methylazetidin-3-ol hydrochloride 97%

Supplier: Ambeed
Warning

A176693-100G A176693-10G A176693-250MG A176693-25G A176693-5G A176693-1G
103807-164EA 9 USD
103807-164 103845-490 76813-386 76813-388 76813-390 76813-392 77082-838 77301-466
3-Methylazetidin-3-ol hydrochloride 97%
3-Methylazetidin-3-ol hydrochloride

Precaución: for Research Use Only

Formula: C4H10ClNO
MW: 123.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11100994
CAS Number: 124668-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR