3-Methyl-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-7-azaindole

A136577-100MG A136577-250MG A136577-1G A136577-5G
103802-324EA 47 USD
103802-324 77272-282 77272-284 77315-234 77315-236
3-Methyl-1H-pyrrolo[2,3-b]pyridine 98%
3-Methyl-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.163 g/mol
Boiling Pt: 284.9 °C (760 mm Hg)
Flash Pt: 127.264 °C
Storage Temperature: Ambient
MDL Number: MFCD08272239
CAS Number: 5654-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR