3-Methyl-6-nitro-1H-indazole 97%
Supplier: Ambeed
|
Warning
|
A210412-100G
A210412-10G
A210412-5G
103848-696EA
10
USD
103848-696
76975-518
76975-520
77087-072
3-Methyl-6-nitro-1H-indazole 97%
3-Methyl-6-nitro-1H-indazole
Precaución: for Research Use Only
Formula:
C₈H₇N₃O₂ MW: 177.1600 g/mol Boiling Pt: 225.2 °C (760 mm Hg) Melting Pt: 187…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD07082704 CAS Number: 6494-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...