3-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3H-imidazo[4,5-b]pyridine 97%

Supplier: Ambeed
Warning

A244577-250MG A244577-1G A244577-5G A244577-100MG
103811-588EA 99 USD
103811-588 103851-420 77000-528 77090-630 77304-324 77600-980
3-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3H-imidazo[4,5-b]pyridine 97%
3-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₈BN₃O₂
MW: 259.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11857689
CAS Number: 1257554-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR