2-Hydroxy-5-nitro-3-picoline 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-5-nitropyridin-2-ol, 3-Methyl-5-nitropyridin-2(1H)-one, 3-Methyl-5-nitro-2-pyridone, 2-Hydroxy-3-methyl-5-nitropyridine

A176974-1G A176974-25G A176974-5G A176974-100G A176974-10G
103807-192EA 5 USD
103807-192 103845-518 77082-886 77082-888 77082-890 77111-258 77111-260
2-Hydroxy-5-nitro-3-picoline 97%
2-Hydroxy-5-nitro-3-picoline

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Boiling Pt: 327.2 °C (760 mm Hg)
Melting Pt: 230…234 °C
Flash Pt: 151.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095073
CAS Number: 21901-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR