3-Methyl-5-nitro-1H-indazole 97%

Supplier: Ambeed
Warning

A207766-100MG A207766-10G A207766-250MG A207766-1G A207766-5G
103817-822EA 13 USD
103817-822 103848-506 77275-042 77275-044 77275-046 77303-044 77303-046
3-Methyl-5-nitro-1H-indazole 97%
3-Methyl-5-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
Boiling Pt: 384.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07082699
CAS Number: 40621-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR