3-Methyl-5-(trifluoromethyl)-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-5-(trifluoromethyl)pyrazole

A301089-1G A301089-25G A301089-5G A301089-100G A301089-100MG A301089-10G A301089-250MG
103814-296EA 8 USD
103814-296 103835-446 76838-122 76838-124 76838-126 77014-948 77014-950 77014-952 77014-954
3-Methyl-5-(trifluoromethyl)-1H-pyrazole 98%
3-Methyl-5-(trifluoromethyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₅F₃N₂
MW: 150.104 g/mol
Melting Pt: 88…90 °C
Flash Pt: 67.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00829309
CAS Number: 10010-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR