3-Methyl-4-nitroanisole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-2-methyl-1-nitrobenzene, 5-Methoxy-2-nitrotoluene, 1-Methoxy-3-methyl-4-nitrobenzene, Methyl 4-nitro-m-tolyl ether

A245945-25G A245945-100G A245945-500G A245945-5G
103851-506EA 7 USD
103851-506 77090-780 77249-876 77249-878 77304-372
3-Methyl-4-nitroanisole 97%
3-Methyl-4-nitroanisole

Precaución: for Research Use Only

Formula: C₈H₉NO₃
Boiling Pt: 280.8 °C (760 mm Hg)
Melting Pt: 48…50 °C
Flash Pt: 135.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00007167
CAS Number: 5367-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR