3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-1H-pyrazole-4-boronic acid, pinacol ester

A115819-10G A115819-250MG A115819-25G A115819-1G A115819-5G A115819-100G
103798-628EA 8 USD
103798-628 103824-488 77026-088 77026-090 77026-092 77311-314 77311-316 77871-482
3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C10H17BN2O2
Boiling Pt: 350.3 °C (760 mm Hg)
Melting Pt: 129…134 °C
Flash Pt: 165.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08690235
CAS Number: 936250-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR