3-Methyl-3-oxetanemethanol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-3-oxethanemethanol

A227846-100G A227846-10G A227846-500G A227846-5G A227846-1000G A227846-25G
103809-618EA 217 USD
103809-618 76987-208 76987-210 76987-212 76987-214 77089-032 77089-034
3-Methyl-3-oxetanemethanol 97%
3-Methyl-3-oxetanemethanol

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
Flash Pt: 97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010273
CAS Number: 3143-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR