3-Methyl-3,8-diazabicyclo[3.2.1]octane dihydrochloride 95%

Supplier: Ambeed
Warning

A224234-1G A224234-100MG A224234-250MG
103819-092EA 88 USD
103819-092 103849-982 77088-624 77165-294 77165-296
3-Methyl-3,8-diazabicyclo[3.2.1]octane dihydrochloride 95%
3-Methyl-3,8-diazabicyclo[3.2.1]octane dihydrochloride

Precaución: for Research Use Only

Formula: C7H16Cl2N2
MW: 199.1213 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18083726
CAS Number: 52407-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR