2-Nitro-m-toluidine 98%

Supplier: Ambeed
Warning

A255886-1G A255886-25G A255886-5G A255886-250MG A255886-10G
103812-222EA 6 USD
103812-222 103852-082 77091-550 77091-552 77091-554 77276-422 77593-600
2-Nitro-m-toluidine 98%
2-Nitro-m-toluidine

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
Flash Pt: 128.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00074851
CAS Number: 601-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR