3-Methyl-2-cyclopenten-1-one 98% stabilized

Supplier: Ambeed
Warning

Synonyms: 3-Methylcyclopent-2-enone, 3-Methyl-2-cyclopentenone

Stabilization: Stabilised with hydroquinone (HQ) 0.1 %

A404807-1G A404807-25G A404807-5G A404807-100G A404807-10G
103827-110EA 9 USD
103827-110 103838-150 76841-632 76841-634 76841-636 77278-802 77278-804
3-Methyl-2-cyclopenten-1-one 98% stabilized
3-Methyl-2-cyclopenten-1-one

Precaución: for Research Use Only

Formula: C₆H₈O
MW: 96.13 g/mol
Boiling Pt: 157.5 °C (760 mm Hg)
Melting Pt: 3…5 °C
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001403
CAS Number: 2758-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR