3-Methyl-1H-pyrazolo[3,4-b]pyridine 98%

Supplier: Ambeed
Warning

A302328-1G A302328-10G
103814-336EA 20 USD
103814-336 103835-492 76838-178 77239-268
3-Methyl-1H-pyrazolo[3,4-b]pyridine 98%
3-Methyl-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C7H7N3
MW: 133.1500 g/mol
Boiling Pt: 304.3 °C (760 mm Hg)
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD09607943
CAS Number: 116834-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR