3-Methyl-1H-pyrazolo[3,4-b]pyridin-5-amine 95%

Supplier: Ambeed
Warning

A216988-100MG A216988-250MG
103818-594EA 104 USD
103818-594 103849-320 76975-732 77671-940
3-Methyl-1H-pyrazolo[3,4-b]pyridin-5-amine 95%
3-Methyl-1H-pyrazolo[3,4-b]pyridin-5-amine

Precaución: for Research Use Only

Formula: C₇H₈N₄
MW: 148.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13193614
CAS Number: 1186608-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR