3-Methyl-1H-pyrazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A763073-1G A763073-10G A763073-25G A763073-5G
103840-898EA 40 USD
103840-898 76845-126 77171-148 77308-278 77308-280
3-Methyl-1H-pyrazole-4-carboxylic acid 97%
3-Methyl-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C5H6N2O2
MW: 126.1133 g/mol
Boiling Pt: 384.2 °C (760 mm Hg)
Melting Pt: 225…230 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00608153
CAS Number: 40704-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR