3-Methyl-1H-pyrazole-4-carbaldehyde 98%

Supplier: Ambeed
Warning

A134380-100G A134380-250MG A134380-1G A134380-25G A134380-5G
103802-012EA 10 USD
103802-012 77272-142 77272-144 77314-762 77314-764 77314-766
3-Methyl-1H-pyrazole-4-carbaldehyde 98%
3-Methyl-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C5H6N2O
Boiling Pt: 295.9 °C (760 mm Hg)
Melting Pt: 107…108.5 °C
Flash Pt: 137.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01313822
CAS Number: 112758-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR