3-Methylindole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Skatol, 3-Methyl-1H-indole, β-Methylindole, Skatole
A126618-100G
A126618-10G
A126618-25G
A126618-5G
A126618-500G
76688-432EA
65
USD
76688-432
76688-434
77313-068
77313-070
77599-832
3-Methylindole 98%
3-Methylindole
Precaución: for Research Use Only
Formula:
C₉H₉N Boiling Pt: 265…266 °C (760 mm Hg) Melting Pt: 92…97 °C Flash Pt: 132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005627 CAS Number: 83-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...