3-Methyl-1H-imidazo[4,5-b]pyridin-2(3H)-one 95%

Supplier: Ambeed
Warning

A187432-100MG A187432-250MG A187432-1G A187432-5G
103816-010EA 83 USD
103816-010 103846-600 76825-118 76825-120 77084-288 77084-290
3-Methyl-1H-imidazo[4,5-b]pyridin-2(3H)-one 95%
3-Methyl-1H-imidazo[4,5-b]pyridin-2(3H)-one

Precaución: for Research Use Only

Formula: C₇H₇N₃O
MW: 149.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01081498
CAS Number: 21991-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR