Methyl-3-quinolyl ether 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methoxyquinoline

A330348-100MG A330348-250MG A330348-1G
103825-548EA 18 USD
103825-548 103836-508 77032-864 77032-866 77306-408
Methyl-3-quinolyl ether 98%
Methyl 3-quinolyl ether

Precaución: for Research Use Only

Formula: C₁₀H₉NO
Boiling Pt: 280.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD16619425
CAS Number: 6931-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR