3-Methoxypyrrolidine hydrochloride 97%

Supplier: Ambeed
Warning

A198262-1G A198262-25G A198262-5G A198262-250MG
103817-006EA 16 USD
103817-006 103847-632 77085-648 77085-650 77085-652 77274-714
3-Methoxypyrrolidine hydrochloride 97%
3-Methoxypyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C5H12ClNO
MW: 137.61 g/mol
Boiling Pt: 171.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09261403
CAS Number: 136725-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR