3-Methoxy-4-pyridineboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Methoxypyridine-4-boronic acid
A340923-1G
A340923-5G
A340923-100MG
A340923-250MG
A340923-25G
103825-818EA
26
USD
103825-818
103836-796
76839-814
76839-816
76989-442
76989-444
76989-446
3-Methoxy-4-pyridineboronic acid 97%
3-Methoxy-4-pyridineboronic acid
Precaución: for Research Use Only
Formula:
C₆H₈BNO₃ MW: 152.9400 g/mol Boiling Pt: 357.0±52.0 °C (760 mm Hg) Flash Pt: 169.7 °C Storage Temperature: Freezer |
MDL Number:
MFCD06801698 CAS Number: 1008506-24-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...