3-Methoxypyridine 98%

Supplier: Ambeed
Warning

A274716-5G A274716-100G A274716-10G A274716-500G A274716-1G A274716-25G
103813-210EA 5 USD
103813-210 103853-138 77092-968 77276-854 77276-856 77276-858 77305-168 77305-170
3-Methoxypyridine 98%
3-Methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 168 °C (760 mm Hg)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00673022
CAS Number: 7295-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR