3-Methoxypyridin-4-amine 96%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-3-methoxypyridine

A208852-1G A208852-5G A208852-100MG A208852-250MG
103817-892EA 226 USD
103817-892 103848-580 77086-910 77086-912 77287-906 77287-908
3-Methoxypyridin-4-amine 96%
3-Methoxypyridin-4-amine

Precaución: for Research Use Only

Formula: C6H8N2O
MW: 124.14 g/mol
Boiling Pt: 277.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09991938
CAS Number: 52334-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR