2-Amino-3-methoxypyrazine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methoxypyrazin-2-amine

A112847-100G A112847-100MG A112847-10G A112847-250MG A112847-1G A112847-5G A112847-25G
103798-118EA 21 USD
103798-118 103823-954 77245-790 77245-792 77245-794 77245-796 77310-780 77310-782 77745-837
2-Amino-3-methoxypyrazine 98%
2-Amino-3-methoxypyrazine

Precaución: for Research Use Only

Formula: C₅H₇N₃O
MW: 125.1300 g/mol
Boiling Pt: 241 °C (760 mm Hg)
Melting Pt: 84…85 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD05722793
CAS Number: 4774-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR