3-Methoxypropionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methoxypropanoic acid

A110514-100G A110514-10G A110514-25G A110514-1G A110514-5G
103797-678EA 6 USD
103797-678 103823-484 77270-858 77270-860 77270-862 77310-280 77310-282
3-Methoxypropionic acid 98%
3-Methoxypropionic acid

Precaución: for Research Use Only

Formula: C₄H₈O₃
MW: 104.1100 g/mol
Boiling Pt: 236.9 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00042654
CAS Number: 2544-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR