3-Methoxypicolinic acid 97%

Supplier: Ambeed
Warning

A190554-1G A190554-5G A190554-250MG
103816-308EA 50 USD
103816-308 103846-908 77084-716 77084-718 77274-402
3-Methoxypicolinic acid 97%
3-Methoxypicolinic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
Boiling Pt: 314.7 °C (760 mm Hg)
Melting Pt: 159…160 °C
Flash Pt: 144.143 °C
Storage Temperature: Ambient
MDL Number: MFCD07367569
CAS Number: 16478-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR