3-Methoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-Methoxyphenyl)metaboric acid, Dihydroxy(3-methoxyphenyl)borane, 3-Methoxybenzeneboronic acid

A110340-100G A110340-10G A110340-1G A110340-500G A110340-5G A110340-25G
103823-462EA 6 USD
103823-462 77189-026 77189-028 77189-030 77189-032 77283-980 77549-136
3-Methoxyphenylboronic acid 97%
3-Methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₃
MW: 151.96 g/mol
Boiling Pt: 318.6 °C (760 mm Hg)
Melting Pt: 160…163 °C
Flash Pt: 146.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00161359
CAS Number: 10365-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR