3-Methoxydiphenylamine 97%

Supplier: Ambeed
Warning

A109248-100G A109248-10G A109248-25G A109248-5G
103808-018EA 451 USD
103808-018 103823-268 77270-744 77270-746 77270-748 77283-846
3-Methoxydiphenylamine 97%
3-Methoxydiphenylamine

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO
Boiling Pt: 329.8 °C (760 mm Hg)
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00008383
CAS Number: 101-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR