3-Methoxy-N,N-dimethylaniline 95%
Supplier: Ambeed
|
Warning
|
A835043-100G
A835043-25G
A835043-5G
103841-184EA
11
USD
103841-184
77281-618
77308-426
77308-428
3-Methoxy-N,N-dimethylaniline 95%
3-Methoxy-N,N-dimethylaniline
Precaución: for Research Use Only
Formula:
C₉H₁₃NO Boiling Pt: 212.4 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00051779 CAS Number: 15799-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...