3-Methoxyisonicotinic acid 96%

Supplier: Ambeed
Warning

A256755-10G A256755-250MG A256755-1G
103812-284EA 14 USD
103812-284 103852-148 76771-798 76771-800 77091-650
3-Methoxyisonicotinic acid 96%
3-Methoxyisonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
Boiling Pt: 399.5 °C (760 mm Hg)
Flash Pt: 195.4 °C
Storage Temperature: Ambient
MDL Number: MFCD07368621
CAS Number: 654663-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR