3-Methoxyisonicotinaldehyde 97%

Supplier: Ambeed
Warning

A284636-1G A284636-100MG A284636-250MG
103813-646EA 190 USD
103813-646 103834-778 76837-304 77277-030 77277-032
3-Methoxyisonicotinaldehyde 97%
3-Methoxyisonicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₇NO₂
Boiling Pt: 262.9 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD04112484
CAS Number: 1849-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR