m-Anisic acid 98%

Supplier: Ambeed
Warning

A152346-1000G A152346-100G A152346-10G A152346-500G A152346-25G
103842-544EA 5 USD
103842-544 76812-262 76812-264 76812-266 76812-268 77299-942
m-Anisic acid 98%
m-Anisic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00002499
CAS Number: 586-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR