3-Methoxy-o-phenylenediamine 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
3-Methoxybenzene-1,2-diamine
A740495-100MG
A740495-250MG
A740495-1G
A740495-25G
A740495-5G
A740495-10G
103840-822EA
275
USD
103840-822
76791-252
76791-254
76845-054
76845-056
76845-058
77873-172
3-Methoxy-o-phenylenediamine 97%
3-Methoxy-o-phenylenediamine
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂O Melting Pt: 39…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD08276903 CAS Number: 37466-89-0 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...