3-Methoxyazetidine hydrochloride 97%

Supplier: Ambeed
Warning

A133003-100G A133003-10G A133003-250MG A133003-1G A133003-25G A133003-5G
103801-814EA 6 USD
103801-814 76983-824 76983-826 76983-828 77314-406 77314-408 77314-410
3-Methoxyazetidine hydrochloride 97%
3-Methoxyazetidine hydrochloride

Precaución: for Research Use Only

Formula: C4H10ClNO
MW: 123.58 g/mol
Boiling Pt: 146.3 °C (760 mm Hg)
Melting Pt: 143…149 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06804514
CAS Number: 148644-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR