3-Methoxy-5-methylpyrazin-2-amine 95%

Supplier: Ambeed
Warning

A113532-1G A113532-100MG
103798-246EA 68 USD
103798-246 103824-086 77297-248 77683-034
3-Methoxy-5-methylpyrazin-2-amine 95%
3-Methoxy-5-methylpyrazin-2-amine

Precaución: For research use only.

Formula: C₆H₉N₃O
MW: 139.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09750038
CAS Number: 89464-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR