3-Methoxy-5-methylaniline 95%

Supplier: Ambeed
Warning

A133414-10G A133414-25G A133414-1G A133414-250MG
103801-872EA 31 USD
103801-872 77027-178 77298-630 77314-518 77871-628
3-Methoxy-5-methylaniline 95%
3-Methoxy-5-methylaniline

Precaución: for Research Use Only

Formula: C8H11NO
Flash Pt: 120.2 °C
Storage Temperature: Ambient
MDL Number: MFCD11036303
CAS Number: 66584-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR