3-Methoxy-4-nitroaniline 98%

Supplier: Ambeed
Warning

A218754-10G A218754-1G A218754-25G
103818-742EA 26 USD
103818-742 103849-612 76826-098 77088-140 77088-142
3-Methoxy-4-nitroaniline 98%
3-Methoxy-4-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
Boiling Pt: 367.2±22.0 °C (760 mm Hg)
Flash Pt: 175.9 °C
Storage Temperature: Ambient
MDL Number: MFCD06661773
CAS Number: 16292-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR