3-Methoxy-4-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A138852-10G A138852-250MG A138852-1G A138852-5G A138852-25G
103802-672EA 21 USD
103802-672 77272-380 77272-382 77299-028 77299-030 77315-732
3-Methoxy-4-methylbenzaldehyde 98%
3-Methoxy-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C9H10O2
MW: 150.1700 g/mol
Boiling Pt: 246.2 °C (760 mm Hg)
Melting Pt: 40.5…41.5 °C
Flash Pt: 105.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114050
CAS Number: 24973-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR