3-Methoxy-p-toluidine 98%

Supplier: Ambeed
Warning

A235945-100G A235945-250MG A235945-1G A235945-25G A235945-5G A235945-10G
103811-054EA 8 USD
103811-054 103850-824 76771-270 76771-272 77089-850 77089-852 77089-854 77877-790
3-Methoxy-p-toluidine 98%
3-Methoxy-p-toluidine

Precaución: for Research Use Only

Formula: C₈H₁₁NO
Boiling Pt: 251 °C (760 mm Hg)
Melting Pt: 56…60 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00025371
CAS Number: 16452-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR