3-Methoxy-4-(1,3-oxazol-5-yl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methoxy-4-(oxazol-5-yl)aniline

A165673-100MG A165673-250MG A165673-1G A165673-10G
103805-986EA 69 USD
103805-986 103844-242 77273-526 77273-528 77300-794 77550-052
3-Methoxy-4-(1,3-oxazol-5-yl)aniline 98%
3-Methoxy-4-(1,3-oxazol-5-yl)aniline

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.199 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03844645
CAS Number: 198821-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR