3-Methoxy-4-(difluoromethoxy)benzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Difluoromethoxy)-3-methoxybenzaldehyde

A140493-10G A140493-25G A140493-1G A140493-5G
103802-874EA 7 USD
103802-874 77258-770 77258-772 77315-986 77315-988
3-Methoxy-4-(difluoromethoxy)benzaldehyde 97%
3-Methoxy-4-(difluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C9H8F2O3
MW: 202.1548 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03412217
CAS Number: 162401-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR